Assay Details
| Gene Symbol: LOC4331253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770982.2 | XP_015626468.1 | 10 - 11 | 1770 | 69 | |
| XM_015770983.2 | XP_015626469.1 | 10 - 11 | 1760 | 69 | ||
| GenBank mRNA | AB081951.1 | - | 10 - 11 | 1587 | 69 | |
| AK103279.1 | - | 10 - 11 | 1736 | 69 | ||
More Information
Set Membership: |
|

LOC4331253
Genomic Map