Assay Details
| Gene Symbol: LOC4331253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770982.2 | XP_015626468.1 | 17 - 18 | 2594 | 103 | |
| XM_015770983.2 | XP_015626469.1 | 17 - 18 | 2584 | 103 | ||
| GenBank mRNA | AB081951.1 | - | 17 - 18 | 2411 | 103 | |
| AK103279.1 | - | 17 - 18 | 2560 | 103 | ||
More Information
Set Membership: |
|

LOC4331253
Genomic Map