Assay Details
| Gene Symbol: LOC4331462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776517.2 | XP_015632003.1 | 10 - 11 | 1201 | 92 | |
| GenBank mRNA | AK065004.1 | - | 10 - 11 | 1195 | 92 | |
| AK121790.1 | - | 10 - 11 | 1195 | 92 | ||
More Information
Set Membership: |
|

LOC4331462
Genomic Map