Assay Details
| Gene Symbol: LOC4331654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774399.2 | XP_015629885.1 | 15 - 16 | 2386 | 84 | |
| GenBank mRNA | AK288414.1 | - | 15 - 16 | 2272 | 84 | |
More Information
Set Membership: |
|

LOC4331654
Genomic Map