Assay Details
| Gene Symbol: LOC4331669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776982.2 | XP_015632468.1 | 3 - 4 | 1290 | 85 | |
| XM_015776984.2 | XP_015632470.1 | 3 - 4 | 1287 | 85 | ||
| XM_015776985.2 | XP_015632471.1 | 3 - 4 | 1245 | 85 | ||
| GenBank mRNA | AK103466.1 | - | 3 - 4 | 1261 | 85 | |
More Information
Set Membership: |
|

LOC4331669
Genomic Map