Assay Details
| Gene Symbol: LOC4331669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776982.2 | XP_015632468.1 | 4 - 5 | 1374 | 91 | |
| XM_015776984.2 | XP_015632470.1 | 4 - 5 | 1371 | 91 | ||
| XM_015776985.2 | XP_015632471.1 | 4 - 5 | 1329 | 91 | ||
| GenBank mRNA | AK103466.1 | - | 4 - 5 | 1345 | 91 | |
More Information
Set Membership: |
|

LOC4331669
Genomic Map