Assay Details
| Gene Symbol: LOC4331669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776982.2 | XP_015632468.1 | 5 - 6 | 1455 | 84 | |
| XM_015776984.2 | XP_015632470.1 | 5 - 6 | 1452 | 84 | ||
| XM_015776985.2 | XP_015632471.1 | 5 - 6 | 1410 | 84 | ||
| GenBank mRNA | AK103466.1 | - | 5 - 6 | 1426 | 84 | |
More Information
Set Membership: |
|

LOC4331669
Genomic Map