Assay Details
| Gene Symbol: LOC4332176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774305.2 | XP_015629791.1 | 3 - 4 | 672 | 133 | |
| GenBank mRNA | AK060384.1 | - | 3 - 4 | 625 | 133 | |
| AK070766.1 | - | 3 - 4 | 676 | 133 | ||
More Information
Set Membership: |
|

LOC4332176
Genomic Map