Assay Details
| Gene Symbol: LOC4332216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777426.2 | XP_015632912.1 | 3 - 4 | 486 | 67 | |
| XM_015777427.2 | XP_015632913.1 | 3 - 4 | 442 | 67 | ||
| XM_015777428.2 | XP_015632914.1 | 3 - 4 | 543 | 67 | ||
More Information
Set Membership: |
|

LOC4332216
Genomic Map