Assay Details
| Gene Symbol: LOC4332243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777550.2 | XP_015633036.1 | 5 - 6 | 736 | 116 | |
| GenBank mRNA | AK100847.1 | - | 5 - 6 | 743 | 116 | |
More Information
Set Membership: |
|

LOC4332243
Genomic Map