Assay Details
| Gene Symbol: LOC4332345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777652.2 | XP_015633138.1 | 4 - 5 | 908 | 83 | |
| GenBank mRNA | AK105267.1 | - | 4 - 5 | 908 | 83 | |
More Information
Set Membership: |
|

LOC4332345
Genomic Map