Assay Details
| Gene Symbol: LOC4332519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776390.2 | XP_015631876.1 | 1 - 2 | 699 | 64 | |
| GenBank mRNA | AK101594.1 | - | 1 - 2 | 685 | 64 | |
| AK119500.1 | - | 1 - 2 | 694 | 64 | ||
| EU267985.1 | - | 1 - 2 | 547 | 64 | ||
More Information
Set Membership: |
|

LOC4332519
Genomic Map