Assay Details
| Gene Symbol: LOC4332537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776563.2 | XP_015632049.1 | 1 - 2 | 417 | 53 | |
| XM_015776564.2 | XP_015632050.1 | 1 - 2 | 417 | 53 | ||
| XM_015776566.2 | XP_015632052.1 | 1 - 2 | 387 | 53 | ||
| GenBank mRNA | AK100042.1 | - | 1 - 2 | 388 | 53 | |
More Information
Set Membership: |
|

LOC4332537
Genomic Map