Assay Details
| Gene Symbol: LOC4332537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776563.2 | XP_015632049.1 | 3 - 4 | 686 | 75 | |
| XM_015776564.2 | XP_015632050.1 | 3 - 4 | 686 | 75 | ||
| XM_015776566.2 | XP_015632052.1 | 3 - 4 | 656 | 75 | ||
| GenBank mRNA | AK100042.1 | - | 3 - 4 | 657 | 75 | |
More Information
Set Membership: |
|

LOC4332537
Genomic Map