Assay Details
| Gene Symbol: LOC4332592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777441.2 | XP_015632927.1 | 1 - 2 | 593 | 121 | |
| GenBank mRNA | AK243161.1 | - | 1 - 2 | 326 | 121 | |
More Information
Set Membership: |
|

LOC4332592
Genomic Map