Assay Details
| Gene Symbol: LOC4332592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777441.2 | XP_015632927.1 | 2 - 3 | 1176 | 66 | |
| GenBank mRNA | AK243161.1 | - | 2 - 3 | 909 | 66 | |
More Information
Set Membership: |
|

LOC4332592
Genomic Map