Assay Details
Gene Symbol: LOC4332724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772383.2 | XP_015627869.1 | 5 - 6 | 1669 | 99 | |
GenBank mRNA | AK102748.1 | - | 5 - 6 | 1565 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
