Assay Details
| Gene Symbol: LOC4332806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777553.2 | XP_015633039.1 | 9 - 10 | 734 | 94 | |
| GenBank mRNA | AK061903.1 | - | 9 - 10 | 635 | 94 | |
| AK065547.1 | - | 9 - 10 | 700 | 94 | ||
More Information
Set Membership: |
|

LOC4332806
Genomic Map