Assay Details
| Gene Symbol: LOC4332811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772702.2 | XP_015628188.1 | 1 - 2 | 409 | 67 | |
| XM_015772703.2 | XP_015628189.1 | 1 - 2 | 411 | 67 | ||
| GenBank mRNA | AK120928.1 | - | 3102 | 67 | ||
More Information
Set Membership: |
|

LOC4332811
Genomic Map