Assay Details
| Gene Symbol: LOC4332811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772702.2 | XP_015628188.1 | 7 - 8 | 921 | 82 | |
| XM_015772703.2 | XP_015628189.1 | 7 - 8 | 923 | 82 | ||
| GenBank mRNA | AK120928.1 | - | 3614 | 82 | ||
More Information
Set Membership: |
|

LOC4332811
Genomic Map