Assay Details
| Gene Symbol: LOC4332976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774692.2 | XP_015630178.1 | 2 - 3 | 325 | 126 | |
| GenBank mRNA | AK069332.1 | - | 2 - 3 | 321 | 126 | |
More Information
Set Membership: |
|

LOC4332976
Genomic Map