Assay Details
| Gene Symbol: LOC4333062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774357.2 | XP_015629843.1 | 12 - 13 | 1881 | 61 | |
| XM_015774358.2 | XP_015629844.1 | 12 - 13 | 2012 | 61 | ||
| GenBank mRNA | AK061713.1 | - | 1 - 2 | 198 | 61 | |
| AK100334.1 | - | 11 - 12 | 1734 | 61 | ||
| HQ895719.1 | - | 11 - 12 | 1699 | 61 | ||
More Information
Set Membership: |
|

LOC4333062
Genomic Map