Assay Details
| Gene Symbol: LOC4333062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774357.2 | XP_015629843.1 | 2 - 3 | 292 | 65 | |
| XM_015774358.2 | XP_015629844.1 | 2 - 3 | 423 | 65 | ||
| GenBank mRNA | AK100334.1 | - | 1 - 2 | 145 | 65 | |
| HQ895719.1 | - | 1 - 2 | 110 | 65 | ||
More Information
Set Membership: |
|

LOC4333062
Genomic Map