Assay Details
| Gene Symbol: LOC4333062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774357.2 | XP_015629843.1 | 4 - 5 | 576 | 76 | |
| XM_015774358.2 | XP_015629844.1 | 4 - 5 | 707 | 76 | ||
| GenBank mRNA | AK100334.1 | - | 3 - 4 | 429 | 76 | |
| HQ895719.1 | - | 3 - 4 | 394 | 76 | ||
More Information
Set Membership: |
|

LOC4333062
Genomic Map