Assay Details
| Gene Symbol: LOC4333062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774357.2 | XP_015629843.1 | 6 - 7 | 883 | 63 | |
| XM_015774358.2 | XP_015629844.1 | 6 - 7 | 1014 | 63 | ||
| GenBank mRNA | AK100334.1 | - | 5 - 6 | 736 | 63 | |
| HQ895719.1 | - | 5 - 6 | 701 | 63 | ||
More Information
Set Membership: |
|

LOC4333062
Genomic Map