Assay Details
| Gene Symbol: LOC4333062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774357.2 | XP_015629843.1 | 10 - 11 | 1495 | 99 | |
| XM_015774358.2 | XP_015629844.1 | 10 - 11 | 1626 | 99 | ||
| GenBank mRNA | AK100334.1 | - | 9 - 10 | 1348 | 99 | |
| HQ895719.1 | - | 9 - 10 | 1313 | 99 | ||
More Information
Set Membership: |
|

LOC4333062
Genomic Map