Assay Details
| Gene Symbol: LOC4333096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773146.2 | XP_015628632.1 | 2 - 3 | 194 | 85 | |
| GenBank mRNA | AK058696.1 | - | 2 - 3 | 172 | 85 | |
| AK071057.1 | - | 2 - 3 | 166 | 85 | ||
More Information
Set Membership: |
|

LOC4333096
Genomic Map