Assay Details
| Gene Symbol: LOC4333162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774946.2 | XP_015630432.1 | 1 - 2 | 307 | 91 | |
| XM_015774948.2 | XP_015630434.1 | 1 - 2 | 307 | 91 | ||
| GenBank mRNA | AK069737.1 | - | 1 - 2 | 281 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774946.2, XM_015774948.2 transcripts.
Set Membership: |
|

LOC4333162
Genomic Map