Assay Details
Gene Symbol: LOC4333162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774946.2 | XP_015630432.1 | 2 - 3 | 1143 | 75 | |
XM_015774947.2 | XP_015630433.1 | 2 - 3 | 1128 | 75 | ||
GenBank mRNA | AK069737.1 | - | 2 - 3 | 1117 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
