Assay Details
| Gene Symbol: LOC4333171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775365.1 | XP_015630851.1 | 13 - 14 | 4043 | 101 | |
| XM_015775366.2 | XP_015630852.1 | 13 - 14 | 4021 | 101 | ||
| GenBank mRNA | AK065099.1 | - | 13 - 14 | 3872 | 101 | |
| AK073276.1 | - | 6 - 7 | 1222 | 101 | ||
| AK103000.1 | - | 10 - 11 | 3478 | 101 | ||
More Information
Set Membership: |
|

LOC4333171
Genomic Map