Assay Details
Gene Symbol: LOC4333171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775365.1 | XP_015630851.1 | 2 - 3 | 422 | 75 | |
GenBank mRNA | AK065099.1 | - | 2 - 3 | 251 | 75 | |
AK067814.1 | - | 2 - 3 | 372 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
