Assay Details
| Gene Symbol: LOC4333678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772513.2 | XP_015627999.1 | 3 - 4 | 565 | 101 | |
| XM_015772514.2 | XP_015628000.1 | 3 - 4 | 565 | 101 | ||
| GenBank mRNA | AK067769.1 | - | 3 - 4 | 568 | 101 | |
More Information
Set Membership: |
|

LOC4333678
Genomic Map