Assay Details
| Gene Symbol: LOC4333741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777825.1 | XP_015633311.1 | 3 - 4 | 742 | 107 | |
| GenBank mRNA | AK100086.1 | - | 3 - 4 | 750 | 107 | |
More Information
Set Membership: |
|

LOC4333741
Genomic Map