Assay Details
| Gene Symbol: LOC4333741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777825.1 | XP_015633311.1 | 4 - 5 | 829 | 108 | |
| GenBank mRNA | AK100086.1 | - | 4 - 5 | 837 | 108 | |
More Information
Set Membership: |
|

LOC4333741
Genomic Map