Assay Details
| Gene Symbol: LOC4333881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777593.2 | XP_015633079.1 | 12 - 13 | 1627 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4333881| Gene Symbol: LOC4333881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777593.2 | XP_015633079.1 | 12 - 13 | 1627 | 76 | |
Set Membership: |
|