Assay Details
| Gene Symbol: LOC4334020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776532.2 | XP_015632018.1 | 14 - 15 | 1663 | 78 | |
| GenBank mRNA | AK069296.1 | - | 14 - 15 | 1621 | 78 | |
| AK105092.1 | - | 2 - 3 | 175 | 78 | ||
| KT199366.1 | - | 14 - 15 | 1611 | 78 | ||
More Information
Set Membership: |
|

LOC4334020
Genomic Map