Assay Details
| Gene Symbol: LOC4334020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776532.2 | XP_015632018.1 | 5 - 6 | 888 | 67 | |
| GenBank mRNA | AK069296.1 | - | 5 - 6 | 846 | 67 | |
| KT199366.1 | - | 5 - 6 | 836 | 67 | ||
More Information
Set Membership: |
|

LOC4334020
Genomic Map