Assay Details
| Gene Symbol: LOC4334020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776532.2 | XP_015632018.1 | 6 - 7 | 943 | 94 | |
| GenBank mRNA | AK069296.1 | - | 6 - 7 | 901 | 94 | |
| KT199366.1 | - | 6 - 7 | 891 | 94 | ||
More Information
Set Membership: |
|

LOC4334020
Genomic Map