Assay Details
| Gene Symbol: LOC4334023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777738.2 | XP_015633224.1 | 2 - 3 | 388 | 91 | |
| GenBank mRNA | AK071715.1 | - | 2 - 3 | 387 | 91 | |
More Information
Set Membership: |
|

LOC4334023
Genomic Map