Assay Details
| Gene Symbol: LOC4334023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777738.2 | XP_015633224.1 | 5 - 6 | 657 | 90 | |
| GenBank mRNA | AK059602.1 | - | 2 - 3 | 197 | 90 | |
| AK071715.1 | - | 5 - 6 | 656 | 90 | ||
More Information
Set Membership: |
|

LOC4334023
Genomic Map