Assay Details
| Gene Symbol: LOC4334029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774969.2 | XP_015630455.1 | 4 - 5 | 1737 | 73 | |
| GenBank mRNA | AK070408.1 | - | 4 - 5 | 1493 | 73 | |
| AK104929.1 | - | 4 - 5 | 1068 | 73 | ||
More Information
Set Membership: |
|

LOC4334029
Genomic Map