Assay Details
| Gene Symbol: LOC4334035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776283.1 | XP_015631769.1 | 2 - 3 | 467 | 75 | |
| XM_015776284.2 | 2 - 3 | 421 | 75 | |||
| GenBank mRNA | AB110184.1 | - | 2 - 3 | 289 | 75 | |
| AK061742.1 | - | 2 - 3 | 409 | 75 | ||
| AK068510.1 | - | 1 - 2 | 2399 | 75 | ||
| BT131597.1 | - | 2 - 3 | 351 | 75 | ||
| BT132110.1 | - | 2 - 3 | 352 | 75 | ||
More Information
Set Membership: |
|

LOC4334035
Genomic Map