Assay Details
| Gene Symbol: LOC4334035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776283.1 | XP_015631769.1 | 4 - 5 | 616 | 86 | |
| XM_015776284.2 | 4 - 5 | 570 | 86 | |||
| GenBank mRNA | AB110184.1 | - | 4 - 5 | 438 | 86 | |
| AK061742.1 | - | 4 - 5 | 558 | 86 | ||
| AK068510.1 | - | 3 - 4 | 2546 | 86 | ||
| BT131597.1 | - | 4 - 5 | 500 | 86 | ||
| BT132110.1 | - | 4 - 5 | 501 | 86 | ||
More Information
Set Membership: |
|

LOC4334035
Genomic Map