Assay Details
| Gene Symbol: LOC4334035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776283.1 | XP_015631769.1 | 5 - 6 | 683 | 65 | |
| XM_015776284.2 | 5 - 6 | 637 | 65 | |||
| GenBank mRNA | AB110184.1 | - | 5 - 6 | 505 | 65 | |
| AK061742.1 | - | 5 - 6 | 625 | 65 | ||
| AK068510.1 | - | 4 - 5 | 2613 | 65 | ||
| BT131597.1 | - | 5 - 6 | 567 | 65 | ||
| BT132110.1 | - | 5 - 6 | 568 | 65 | ||
More Information
Set Membership: |
|

LOC4334035
Genomic Map