Assay Details
| Gene Symbol: LOC9269802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772503.2 | XP_015627989.1 | 18 - 19 | 1917 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9269802| Gene Symbol: LOC9269802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772503.2 | XP_015627989.1 | 18 - 19 | 1917 | 86 | |
Set Membership: |
|