Assay Details
| Gene Symbol: LOC4335713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780210.2 | XP_015635696.1 | 6 - 7 | 1467 | 84 | |
| GenBank mRNA | AK065158.1 | - | 6 - 7 | 1445 | 84 | |
More Information
Set Membership: |
|

LOC4335713
Genomic Map