Assay Details
| Gene Symbol: LOC4335733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779954.2 | XP_015635440.1 | 3 - 4 | 515 | 145 | |
| GenBank mRNA | AK240901.1 | - | 465 | 145 | ||
More Information
Set Membership: |
|

LOC4335733
Genomic Map