Assay Details
| Gene Symbol: LOC4335746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778740.2 | XP_015634226.1 | 8 - 9 | 883 | 69 | |
| GenBank mRNA | AK107531.1 | - | 8 - 9 | 890 | 69 | |
More Information
Set Membership: |
|

LOC4335746
Genomic Map