Assay Details
| Gene Symbol: LOC4335746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778740.2 | XP_015634226.1 | 9 - 10 | 976 | 58 | |
| GenBank mRNA | AK107531.1 | - | 9 - 10 | 983 | 58 | |
More Information
Set Membership: |
|

LOC4335746
Genomic Map