Assay Details
| Gene Symbol: LOC4334201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772828.2 | XP_015628314.1 | 3 - 4 | 815 | 88 | |
| GenBank mRNA | AK071181.1 | - | 3 - 4 | 689 | 88 | |
| AK240962.1 | - | 1525 | 88 | |||
| AK287934.1 | - | 621 | 88 | |||
More Information
Set Membership: |
|

LOC4334201
Genomic Map